Tisk…

2-Acetyl-7-bromo-5-chloro-2H-indazole

Dodavatel: Apollo Scientific
Varování

APOSOR110361-1GEA 5860 CZK
APOSOR110361-1G
2-Acetyl-7-bromo-5-chloro-2H-indazole
2-Acetyl-7-bromo-5-chloro-2H-indazole
Vzorec: C₉H₆BrClN₂O
MW: 273,514 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619589
CAS číslo: 1355171-38-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR