Tisk…

2-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one

Dodavatel: Apollo Scientific
Varování

APOSOR110388-1GEA 5330 CZK
APOSOR110388-1G
2-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one
2-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one
Vzorec: C₉H₈N₂O₄
MW: 208,171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17168279
CAS číslo: 132522-85-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR