Tisk…

[4-(1,3,4-Oxadiazol-2-yl)phenyl]methanol

Dodavatel: Apollo Scientific
Varování

APOSOR110429-1GEA 6450 CZK
APOSOR110429-1G
[4-(1,3,4-Oxadiazol-2-yl)phenyl]methanol
[4-(1,3,4-Oxadiazol-2-yl)phenyl]methanol
Vzorec: C₉H₈N₂O₂
MW: 176,172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18804155
CAS číslo: 1378699-97-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR