Tisk…

1-(Bromomethyl)-2-(2-isopropylphenoxy)benzene

APOSOR110513-1GEA 0 CZK
APOSOR110513-1G
1-(Bromomethyl)-2-(2-isopropylphenoxy)benzene
1-(Bromomethyl)-2-(2-isopropylphenoxy)benzene
Vzorec: C₁₆H₁₇BrO
MW: 305,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619593
CAS číslo: 1427461-01-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR