Tisk…

1-(Bromomethyl)-2-(3-tert-butylphenoxy)benzene

Dodavatel: Apollo Scientific

APOSOR110515-1GEA 8630 CZK
APOSOR110515-1G
1-(Bromomethyl)-2-(3-tert-butylphenoxy)benzene
1-(Bromomethyl)-2-(3-tert-butylphenoxy)benzene
Vzorec: C₁₇H₁₉BrO
MW: 319,236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619595
CAS číslo: 1427460-79-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR