Tisk…

3-[4-(tert-Butoxycarbonyl)homopiperazin-1-yl]propanoic acid

Dodavatel: Apollo Scientific

APOSOR110607-10GEA 47940 CZK
APOSOR110607-10G APOSOR110607-1G APOSOR110607-5G
3-[4-(tert-Butoxycarbonyl)homopiperazin-1-yl]propanoic acid
3-[4-(tert-Butoxycarbonyl)homopiperazin-1-yl]propanoic acid
Vzorec: C₁₃H₂₄N₂O₄
MW: 272,341 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664951
CAS číslo: 874801-64-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR