Tisk…

Diethyl 5-amino-3-(2-ethoxy-2-oxoethyl)thiophene-2,4-dicarboxylate

Varování

APOSOR110635-1GEA 0 CZK
APOSOR110635-1G
Diethyl 5-amino-3-(2-ethoxy-2-oxoethyl)thiophene-2,4-dicarboxylate
Diethyl 5-amino-3-(2-ethoxy-2-oxoethyl)thiophene-2,4-dicarboxylate
Vzorec: C₁₄H₁₉NO₆S
MW: 329,369 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09837427
CAS číslo: 58168-14-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR