6-(2-Furyl)-2-hydroxy-4-(2-thienyl)nicotinonitrile
Dodavatel: Apollo Scientific
APOSOR110681-1GEA
3340
CZK
APOSOR110681-1G
6-(2-Furyl)-2-hydroxy-4-(2-thienyl)nicotinonitrile
6-(2-Furyl)-2-hydroxy-4-(2-thienyl)nicotinonitrile
Vzorec:
C₁₄H₈N₂O₂S MW: 268,291 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07774476 CAS číslo: 111121-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...