Tisk…

7-Bromo-1-benzothiophene-2-carbohydrazide

Dodavatel: Apollo Scientific
Varování

APOSOR110808-1GEA 2450 CZK
APOSOR110808-1G
7-Bromo-1-benzothiophene-2-carbohydrazide
7-Bromo-1-benzothiophene-2-carbohydrazide
Vzorec: C₉H₇BrN₂OS
MW: 271,134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12032269
CAS číslo: 1171927-51-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR