Tisk…

Ethyl 3-bromo-1H-indazole-5-carboxylate

Dodavatel: Apollo Scientific
Varování

APOSOR110865-1GEA 8140 CZK
APOSOR110865-1G
Ethyl 3-bromo-1H-indazole-5-carboxylate
Ethyl 3-bromo-1H-indazole-5-carboxylate
Vzorec: C₁₀H₉BrN₂O₂
MW: 269,095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21607224
CAS číslo: 192945-25-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR