Tisk…

2-tert-Butylimidazo[1,2-a]pyridine-6-carboxylic acid

Dodavatel: Apollo Scientific

APOSOR110993-1GEA 10290 CZK
APOSOR110993-1G
2-tert-Butylimidazo[1,2-a]pyridine-6-carboxylic acid
2-tert-Butylimidazo[1,2-a]pyridine-6-carboxylic acid
Vzorec: C₁₂H₁₄N₂O₂
MW: 218,252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14689252
CAS číslo: 1248085-24-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR