Tisk…

1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]piperazine

Dodavatel: Apollo Scientific

APOSOR110994-1GEA 10290 CZK
APOSOR110994-1G
1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]piperazine
1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]piperazine
Vzorec: C₉H₁₅N₅O₂
MW: 225,248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16076302
CAS číslo: 1248142-10-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR