Tisk…

4-(2-Methyl-1,3-thiazol-4-yl)benzene-1-sulphonamide

Dodavatel: Apollo Scientific

APOSOR111118-1GEA 8300 CZK
APOSOR111118-1G
4-(2-Methyl-1,3-thiazol-4-yl)benzene-1-sulphonamide
4-(2-Methyl-1,3-thiazol-4-yl)benzene-1-sulphonamide
Vzorec: C₁₀H₁₀N₂O₂S₂
MW: 254,329 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11650716
CAS číslo: 1094460-88-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR