Tisk…

2-Chloro-N-(2,4,6-trimethoxyphenyl)acetamide

Dodavatel: Apollo Scientific

APOSOR111234-1GEA 9660 CZK
APOSOR111234-1G
2-Chloro-N-(2,4,6-trimethoxyphenyl)acetamide
2-Chloro-N-(2,4,6-trimethoxyphenyl)acetamide
Vzorec: C₁₁H₁₄ClNO₄
MW: 259,686 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30188162
CAS číslo: 1245796-20-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR