Tisk…

2-(2-Cyclohexylethyl)-1H-isoindole-1,3(2H)-dione

Dodavatel: Apollo Scientific

APOSOR111353-1GEA 7710 CZK
APOSOR111353-1G APOSOR111353-5G
2-(2-Cyclohexylethyl)-1H-isoindole-1,3(2H)-dione
2-(2-Cyclohexylethyl)-1H-isoindole-1,3(2H)-dione
Vzorec: C₁₆H₁₉NO₂
MW: 257,328 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09923370
CAS číslo: 41763-92-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR