Tisk…

3-(Naphth-1-yl)quinoxaline-5-carboxylic acid

Dodavatel: Apollo Scientific

APOSOR11700-500MGEA 0 CZK
APOSOR11700-500MG APOSOR11700-1G APOSOR11700-50MG
3-(Naphth-1-yl)quinoxaline-5-carboxylic acid
3-(Naphth-1-yl)quinoxaline-5-carboxylic acid
Vzorec: C₁₉H₁₂N₂O₂
MW: 300,311 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660392
CAS číslo: 904818-32-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR