Tetramethyl 5-formyl-5'-methylbis-(2-aminophenoxy-methylene)-N,N,N',N'-tetraacetate
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR1275T-10MGEA
9130
CZK
APOSOR1275T-10MG
Tetramethyl 5-formyl-5'-methylbis-(2-aminophenoxy-methylene)-N,N,N',N'-tetraacetate
Tetramethyl 5-formyl-5'-methylbis-(2-aminophenoxy-methylene)-N,N,N',N'-tetraacetate
A new fluorescent chelating indicator to study the physiological role of cytosolic free Calcium.
Vzorec:
C₂₈H₃₄N₂O₁₁ MW: 574,576 g/mol Bod tání: 132…133 °C Storage Temperature: Freezer |
MDL Number:
MFCD00291363 CAS číslo: 96315-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...