Tisk…

Ethyl pyrrolo[1,2-a]quinoline-2-carboxylate

Dodavatel: Apollo Scientific

APOSOR1373-500MGEA 5720 CZK
APOSOR1373-500MG APOSOR1373-1G
Ethyl pyrrolo[1,2-a]quinoline-2-carboxylate
Ethyl pyrrolo[1,2-a]quinoline-2-carboxylate
Vzorec: C₁₅H₁₃NO₂
MW: 239,269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08056189
CAS číslo: 76577-82-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR