Tisk…

4-Nitro-1,5,6,7-tetrahydro-5,5,7,7-tetramethylcyclopenta[f]indazole

Dodavatel: Apollo Scientific
Varování

APOSOR16155-1GEA 7670 CZK
APOSOR16155-1G
4-Nitro-1,5,6,7-tetrahydro-5,5,7,7-tetramethylcyclopenta[f]indazole
4-Nitro-1,5,6,7-tetrahydro-5,5,7,7-tetramethylcyclopenta[f]indazole
Vzorec: C₁₄H₁₇N₃O₂
MW: 259,304 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109510
CAS číslo: 1209368-38-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR