Tisk…

6-Bromobenzo[c]isothiazole

Dodavatel: Apollo Scientific
Varování

APOSOR16643-500MGEA 0 CZK
APOSOR16643-500MG
6-Bromobenzo[c]isothiazole
6-Bromobenzo[c]isothiazole
Vzorec: C₇H₄BrNS
MW: 214,082 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13176692
CAS číslo: 20712-08-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR