Tisk…

3-Iodo-7-methyl-1H-indole, N-BOC protected 98%

Dodavatel: Apollo Scientific
Varování

APOSOR1735-1GEA 5200 CZK
APOSOR1735-1G APOSOR1735-5G
3-Iodo-7-methyl-1H-indole, N-BOC protected 98%
3-Iodo-7-methyl-1H-indole, N-BOC protected
Vzorec: C₁₄H₁₆INO₂
MW: 357,187 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05864783
CAS číslo: 914349-35-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR