5-Hydroxy-4H-chromen-4-one 95%
Dodavatel: Apollo Scientific
APOSOR21982-1GEA
11410
CZK
APOSOR21982-1G
5-Hydroxy-4H-chromen-4-one 95%
5-Hydroxy-4H-chromen-4-one
Vzorec:
C₉H₆O₃ MW: 162,142 g/mol Bod tání: 127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100605 CAS číslo: 3952-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...