Tisk…

Methyl 3-amino-4-(phenylsulphonyl)thiophene-2-carboxylate

Dodavatel: Apollo Scientific

APOSOR25127-1GEA 15110 CZK
APOSOR25127-1G APOSOR25127-250MG
Methyl 3-amino-4-(phenylsulphonyl)thiophene-2-carboxylate
Methyl 3-amino-4-(phenylsulphonyl)thiophene-2-carboxylate
Vzorec: C₁₂H₁₁NO₄S₂
MW: 297,35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00067908
CAS číslo: 175201-55-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR