Tisk…

3,4-dimethylthieno[2,3-b]thiophene-2,5-dicarboxylic acid 95+%

Dodavatel: Apollo Scientific
Varování

APOSOR25290-250MGEA 4500 CZK
APOSOR25290-250MG
3,4-dimethylthieno[2,3-b]thiophene-2,5-dicarboxylic acid 95+%
3,4-dimethylthieno[2,3-b]thiophene-2,5-dicarboxylic acid
Vzorec: C₁₀H₈O₄S₂
MW: 256,298 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00067989
CAS číslo: 175202-55-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR