Tisk…

1-(Aminomethyl)cyclohexanecarboxylic acid, N-FMOC protected

Dodavatel: Apollo Scientific

APOSOR2667-1GEA 12240 CZK
APOSOR2667-1G APOSOR2667-250MG
1-(Aminomethyl)cyclohexanecarboxylic acid, N-FMOC protected
1-(Aminomethyl)cyclohexanecarboxylic acid, N-FMOC protected
Vzorec: C₂₃H₂₅NO₄
MW: 379,449 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264208
CAS číslo: 220145-22-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR