Tisk…

1-Chloroisoquinoline-8-carboxylic acid

Dodavatel: Apollo Scientific
Varování

APOSOR300081-500MGEA 0 CZK
APOSOR300081-500MG
1-Chloroisoquinoline-8-carboxylic acid
1-Chloroisoquinoline-8-carboxylic acid
Vzorec: C₁₀H₆ClNO₂
MW: 207,613 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21365098
CAS číslo: 1337879-52-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR