Tisk…

6-(4-Morpholinylmethyl)-3-pyridinyl boronic acid

Dodavatel: Apollo Scientific

APOSOR301056-1GEA 41660 CZK
APOSOR301056-1G
6-(4-Morpholinylmethyl)-3-pyridinyl boronic acid
6-(4-Morpholinylmethyl)-3-pyridinyl boronic acid
Vzorec: C₁₀H₁₅BN₂O₃
MW: 222,049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18260451
CAS číslo: 934505-32-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR