Tisk…

N-(6-Aminopyrimidin-4-yl)acetamide

Dodavatel: Apollo Scientific

APOSOR302041-1GEA 48270 CZK
APOSOR302041-1G
N-(6-Aminopyrimidin-4-yl)acetamide
N-(6-Aminopyrimidin-4-yl)acetamide
Vzorec: C₆H₈N₄O
MW: 152,154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01689943
CAS číslo: 89533-23-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR