Tisk…

6-Benzyl-2,4-dihydroxy-5H-pyrrolo[3,4-d]pyrimidin-7(6H)-one

Dodavatel: Apollo Scientific

APOSOR302140-1GEA 26750 CZK
APOSOR302140-1G
6-Benzyl-2,4-dihydroxy-5H-pyrrolo[3,4-d]pyrimidin-7(6H)-one
6-Benzyl-2,4-dihydroxy-5H-pyrrolo[3,4-d]pyrimidin-7(6H)-one
Vzorec: C₁₃H₁₁N₃O₃
MW: 257,245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23704679
CAS číslo: 19068-66-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR