Tisk…

Pyridine-2,4-diyldimethanol

Dodavatel: Apollo Scientific

APOSOR302209-1GEA 24870 CZK
APOSOR302209-1G
Pyridine-2,4-diyldimethanol
Pyridine-2,4-diyldimethanol
Vzorec: C₇H₉NO₂
MW: 139,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617173
CAS číslo: 21071-04-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR