Tisk…

6-Bromo-1H-pyrrolo[2,3-b]pyridin-4-amine

Dodavatel: Apollo Scientific

APOSOR302356-1GEA 26020 CZK
APOSOR302356-1G
6-Bromo-1H-pyrrolo[2,3-b]pyridin-4-amine
6-Bromo-1H-pyrrolo[2,3-b]pyridin-4-amine
Vzorec: C₇H₆BrN₃
MW: 212,047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09880154
CAS číslo: 1000340-72-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR