Tisk…

3-Azabicyclo[3.3.1]nonane hydrochloride

Dodavatel: Apollo Scientific

APOSOR302421-1GEA 0 CZK
APOSOR302421-1G
3-Azabicyclo[3.3.1]nonane hydrochloride
3-Azabicyclo[3.3.1]nonane hydrochloride
Vzorec: C₈H₁₅N·ClH
MW: 161,672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20544769
CAS číslo: 60364-11-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR