Tisk…

2-Ethynylpiperidine hydrochloride

Dodavatel: Apollo Scientific

APOSOR302454-1GEA 36040 CZK
APOSOR302454-1G
2-Ethynylpiperidine hydrochloride
2-Ethynylpiperidine hydrochloride
Vzorec: C₇H₁₁N·ClH
MW: 145,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19689453
CAS číslo: 1380680-53-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR