Tisk…

2-[4-(2-Methoxyphenyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Dodavatel: Apollo Scientific
Varování

APOSOR303544-500MGEA 3910 CZK
APOSOR303544-500MG APOSOR303544-1G APOSOR303544-5G
2-[4-(2-Methoxyphenyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-[4-(2-Methoxyphenyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Vzorec: C₁₉H₂₃BO₃
MW: 310,195 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22493566
CAS číslo: 881314-65-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR