Tisk…

Methyl (2S)-1-(2-methoxy-2-oxoacetyl)pyrrolidine-2-carboxylate

Dodavatel: Apollo Scientific

APOSOR303972-500MGEA 5230 CZK
APOSOR303972-500MG APOSOR303972-5G APOSOR303972-1G
Methyl (2S)-1-(2-methoxy-2-oxoacetyl)pyrrolidine-2-carboxylate
Methyl (2S)-1-(2-methoxy-2-oxoacetyl)pyrrolidine-2-carboxylate
Vzorec: C₉H₁₃NO₅
MW: 215,203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179272
CAS číslo: 139419-63-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR