Tisk…

3-(Benzyloxy)-2,2-dimethylcyclobutan-1-one

Dodavatel: Apollo Scientific

APOSOR304375-1GEA 32730 CZK
APOSOR304375-1G
3-(Benzyloxy)-2,2-dimethylcyclobutan-1-one
3-(Benzyloxy)-2,2-dimethylcyclobutan-1-one
Vzorec: C₁₃H₁₆O₂
MW: 204,265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21755679
CAS číslo: 2063-92-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR