Tisk…

2,2-Dimethylnonanoic acid

Dodavatel: Apollo Scientific

Varování

APOSOR4137-1GEA 3840 CZK
APOSOR4137-1G
2,2-Dimethylnonanoic acid
2,2-Dimethylnonanoic acid
Vzorec: C₁₁H₂₂O₂
MW: 186,291 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093530
CAS číslo: 14250-75-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR