Tisk…

1-Bromo-4-ethoxy-2-methylbenzene 97%

Dodavatel: Apollo Scientific

APOSOR42253-5GEA 3510 CZK
APOSOR42253-5G
1-Bromo-4-ethoxy-2-methylbenzene 97%
1-Bromo-4-ethoxy-2-methylbenzene
Vzorec: C₉H₁₁BrO
MW: 215,087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917736
CAS číslo: 68155-69-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR