Tisk…

2-(Aminomethyl)piperidine, 2-BOC protected

Dodavatel: Apollo Scientific
Nebezpečí

APOSOR4459-250MGEA 417 CZK
APOSOR4459-250MG
2-(Aminomethyl)piperidine, 2-BOC protected
2-(Aminomethyl)piperidine, 2-BOC protected
Vzorec: C₁₁H₂₂N₂O₂
MW: 214,305 g/mol
Bod tání: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD02181068
CAS číslo: 141774-61-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR