Tisk…

2-(2,5-Dihydro-3-furanyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Dodavatel: Apollo Scientific

APOSOR450022-100MGEA 7280 CZK
APOSOR450022-100MG APOSOR450022-250MG
2-(2,5-Dihydro-3-furanyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(2,5-Dihydro-3-furanyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Vzorec: C₁₀H₁₇BO₃
MW: 196,051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16996209
CAS číslo: 212127-80-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR