Tisk…

N-(4-Bromophenyl)-1,2,3,4-thiatriazol-5-amine

Dodavatel: Apollo Scientific

APOSOR450123-250MGEA 3940 CZK
APOSOR450123-250MG
N-(4-Bromophenyl)-1,2,3,4-thiatriazol-5-amine
N-(4-Bromophenyl)-1,2,3,4-thiatriazol-5-amine
Vzorec: C₇H₅BrN₄S
MW: 257,11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32264088
CAS číslo: 68371-97-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR