Tisk…

1-(Diphenylmethyl)-2-azetidinemethanol

Dodavatel: Apollo Scientific

APOSOR451014-1GEA 33060 CZK
APOSOR451014-1G
1-(Diphenylmethyl)-2-azetidinemethanol
1-(Diphenylmethyl)-2-azetidinemethanol
Vzorec: C₁₇H₁₉NO
MW: 253,339 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07786954
CAS číslo: 72351-68-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR