(S)-4-Boc-3-(aminomethyl)-morpholine
Dodavatel: Apollo Scientific
APOSOR451063-1GEA
43640
CZK
APOSOR451063-1G
(S)-4-Boc-3-(aminomethyl)-morpholine
(S)-4-Boc-3-(aminomethyl)-morpholine
Vzorec:
C₁₀H₂₀N₂O₃ MW: 216,277 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06796250 CAS číslo: 1187929-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...