1-(4-Bromophenyl)-cyclobutanemethanamine
Dodavatel: Apollo Scientific
APOSOR451072-1GEA
24870
CZK
APOSOR451072-1G
1-(4-Bromophenyl)-cyclobutanemethanamine
1-(4-Bromophenyl)-cyclobutanemethanamine
Vzorec:
C₁₁H₁₄BrN MW: 240,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11188929 CAS číslo: 1039932-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...