Tisk…

N-(Cyclopropylmethyl)-4-methoxy-benzylamine

Dodavatel: Apollo Scientific

APOSOR451087-1GEA 33060 CZK
APOSOR451087-1G
N-(Cyclopropylmethyl)-4-methoxy-benzylamine
N-(Cyclopropylmethyl)-4-methoxy-benzylamine
Vzorec: C₁₂H₁₇NO
MW: 191,269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11140261
CAS číslo: 1019611-33-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR