(E)-3-[4-(tert-Butyl)benzoyl]acrylic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR4543-1GEA
5200
CZK
APOSOR4543-1G
(E)-3-[4-(tert-Butyl)benzoyl]acrylic acid 98%
(E)-3-[4-(tert-Butyl)benzoyl]acrylic acid
Vzorec:
C₁₄H₁₆O₃ MW: 232,275 g/mol Bod tání: 127…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00171556 CAS číslo: 97678-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...