Tisk…

(S)-4-Benzyl-3-morpholinecarboxylic acid ≥98%

Dodavatel: Apollo Scientific

APOSOR470023-100MGEA 4100 CZK
APOSOR470023-100MG
(S)-4-Benzyl-3-morpholinecarboxylic acid ≥98%
(S)-4-Benzyl-3-morpholinecarboxylic acid
Vzorec: C₁₂H₁₅NO₃
MW: 221,252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06799463
CAS číslo: 1235011-96-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR