Tisk…

Ethyl 3-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate

Dodavatel: Apollo Scientific

APOSOR470035-5GEA 10450 CZK
APOSOR470035-5G
Ethyl 3-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate
Ethyl 3-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate
Vzorec: C₁₃H₁₄N₂O₃
MW: 246,262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12198872
CAS číslo: 1166756-76-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR