(R)-1-(1H-Benzimidazol-2-yl)-2-methylpropylamine hydrochloride
Dodavatel: Apollo Scientific
APOSOR470050-5GEA
26090
CZK
APOSOR470050-5G
(R)-1-(1H-Benzimidazol-2-yl)-2-methylpropylamine hydrochloride
(R)-1-(1H-Benzimidazol-2-yl)-2-methylpropylamine hydrochloride
Vzorec:
C₁₁H₁₅N₃·ClH MW: 225,718 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12828691 CAS číslo: 1234863-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...