Ethyl 2-(Methylsulphonyl)nicotinate
Dodavatel: Apollo Scientific
APOSOR470054-5GEA
22920
CZK
APOSOR470054-5G
Ethyl 2-(Methylsulphonyl)nicotinate
Ethyl 2-(Methylsulphonyl)nicotinate
Vzorec:
C₉H₁₁NO₄S MW: 229,253 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12828713 CAS číslo: 1186663-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...